Learn More
Aviva Systems Biology Corp ALG5 Antibody - N-terminal region (ARP61995_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61995_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ALG5. It was validated on Western Blot. . Alternate names: RP11-421P11.2, bA421P11.2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-287-536