Learn More
Aviva Systems Biology Corp Alg8 Antibody - middle region (ARP44532_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44532_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Alg8. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Rat. Alternate names: MGC124750, Alg8ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-285-151