Learn More
Percival Scientific Algae Growth Chamber, AL30L2H11, 9.9 cu. ft.,SciWhite LEDs, 2 tiers, 7-44° C ±0.5°, IntellusUltra controller, touchscreen, advanced additive ultrasonic humidifier
SDP
Catalog No. NC3794953
Encompass_Preferred
The AL30L2H11 algae chamber used for algae with higher light intensity, and short plants w/ moderate light intensity. SciWhite LEDs are dimmable between 10-100% output with up to 440 µmoles/m2/s light irradiance @ 6”. Standard IntellusUltra controller, touchscreen, Advanced additive ultrasonic humidifier. Interior/exterior of smooth white galvanized steel with foamed-in-place non-CFC insulation, stainless-steel floor for corrosion resistance. Conditioned air circulates through rear wall plenum, travels horizontally across each shelf; then vertically upward to unit cooler. Two white epoxy coated steel wire shelves adjustable in ½” increments. Self-contained air-cooled condensing unit with hot gas bypass system for continuous compressor operation extends life, ensuring precise temperature control by alternately cycling refrigerant and hot gas to coil; also prolongs life of compressor, and eliminates risk of ice build up in coil. Contact sales@percival-scientific.com for more options.
Catalog No. NC3794953 Supplier Percival Scientific Supplier No. AL30L2H11
Add to Cart
Add to Cart
Request a Quote
The AL30L2H11 algae chamber used for algae with higher light intensity, and short plants w/ moderate light intensity. SciWhite LEDs are dimmable between 10-100% output with up to 440 µmoles/m2/s light irradiance @ 6”. Standard IntellusUltra controller, touchscreen, Advanced additive ultrasonic humidifier. Interior/exterior of smooth white galvanized steel with foamed-in-place non-CFC insulation, stainless-steel floor for corrosion resistance. Conditioned air circulates through rear wall plenum, travels horizontally across each shelf; then vertically upward to unit cooler. Two white epoxy coated steel wire shelves adjustable in ½” increments. Self-contained air-cooled condensing unit with hot gas bypass system for continuous compressor operation extends life, ensuring precise temperature control by alternately cycling refrigerant and hot gas to coil; also prolongs life of compressor, and eliminates risk of ice build up in coil. Contact sales@percival-scientific.com for more options.