Learn More
Ace Glass, Inc. Distillation system, alginates assay
SDP
Catalog No. NC0652524
Encompass
Distillation system, alginates assay. The system includes a metering valve with flow meter tube set, 250ml flask and mantle, reflux condenser with u-tube and hose connection adapters and a 250ml gas washing bottle. 5/16in ID Vinyl tubing.
Catalog No. NC0652524 Supplier Ace Glass, Inc. Supplier No. 6546155
Distillation system, alginates assay. The system includes a metering valve with flow meter tube set, 250ml flask and mantle, reflux condenser with u-tube and hose connection adapters and a 250ml gas washing bottle. 5/16in ID Vinyl tubing.