Learn More
Research Products International Corp Alginic Acid Sodium Salt, 250 Grams
SDP
Catalog No. 50213156
Encompass_Preferred
From Phaeophyceae (algae). A mixture of polyuronic acids composed of residues of D-mannuronic acid and L-glucuronic acids. Alginates are used as suspending and thickening agents and in the preparation of water miscible gels. Viscosity: 1%, 25 cps.
Catalog No. 50-213-156 Supplier Research Products International Corp Supplier No. A50300250.0
From Phaeophyceae (algae). A mixture of polyuronic acids composed of residues of D-mannuronic acid and L-glucuronic acids. Alginates are used as suspending and thickening agents and in the preparation of water miscible gels. Viscosity: 1%, 25 cps.