Learn More
Alkali Scientific Alginic Acid Sodium Salt, 500 G
SDP
Catalog No. 5000660432
Encompass_Preferred
Alginic Acid Sodium Salt (500g) is a high-viscosity laboratory-grade chemical used in a variety of applications including gelation, thickening, and stabilization. It is especially valuable in the food industry, pharmaceutical formulations, and medical research. Alginic acid’s ability to form gels in the presence of calcium ions makes it useful for controlled drug delivery systems and wound dressings. The 500g quantity is perfect for larger scale experiments or commercial applications where alginate-based formulations are required. This product ensures reliable consistency and performance, and its purity supports accurate results in research and diagnostic applications.
Catalog No. 50-006-60432 Supplier Alkali Scientific Supplier No. ALG003CP
Alginic Acid Sodium Salt (500g) is a high-viscosity laboratory-grade chemical used in a variety of applications including gelation, thickening, and stabilization. It is especially valuable in the food industry, pharmaceutical formulations, and medical research. Alginic acid’s ability to form gels in the presence of calcium ions makes it useful for controlled drug delivery systems and wound dressings. The 500g quantity is perfect for larger scale experiments or commercial applications where alginate-based formulations are required. This product ensures reliable consistency and performance, and its purity supports accurate results in research and diagnostic applications.