Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alginic acid sodium salt, high viscosity

Catalog No. AAJ6188730
Change view
Click to view available options
Quantity:
100 g
250 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6188730 250 g
AAJ6188722 100 g
AAJ61887A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6188730 Supplier Thermo Scientific Chemicals Supplier No. J6188730
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

A gelling polysaccharide extracted from giant brown seaweed, useful in viscosity studies | CAS: 9005-38-3 | (C6H7O7)A(C6H7O7)BNa

A gelling polysaccharide extracted from giant brown seaweed, useful in viscosity studiesAlginic acid sodium salt is widely utilized in food industry as flavorless gum, which enhances viscosity and as an emulsifier. It finds application as thickener for reactive dyestuffs, which is used in textile screen printing and carpet jet-printing. It is used indigestion tablets and the preparation of dental impressions. As a polymer, it combines with chitosan to synthesize a biodegradable porous scaffold, which is useful for tissue engineering applications. It acts as a food additive and involved in the production of gel-like foods such as bakers. It is employed as thickener for reactive dyestuffs in textile screen-printing and carpet jet-printing. It is also used as encapsulating agents of microparticles of beta-galactosidae and in the preparation of alginate hydrogels with tunable mechanical properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A gelling polysaccharide extracted from giant brown seaweed, useful in viscosity studiesAlginic acid sodium salt is widely utilized in food industry as flavorless gum, which enhances viscosity and as an emulsifier. It finds application as thickener for reactive dyestuffs, which is used in textile screen printing and carpet jet-printing. It is used indigestion tablets and the preparation of dental impressions. As a polymer, it combines with chitosan to synthesize a biodegradable porous scaffold, which is useful for tissue engineering applications. It acts as a food additive and involved in the production of gel-like foods such as bakers. It is employed as thickener for reactive dyestuffs in textile screen-printing and carpet jet-printing. It is also used as encapsulating agents of microparticles of beta-galactosidae and in the preparation of alginate hydrogels with tunable mechanical properties.

Solubility
Soluble in water. Insoluble in alcohol, chloroform and ether.

Notes
Incompatible with strong oxidizing agents, acids and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9005-38-3
Molecular Formula (C6H7O7)A(C6H7O7)BNa
MDL Number MFCD00081310
Synonym Algin; Sodium alginate

Specifications

Color White
Quantity 250 g
Sensitivity Hygroscopic
Merck Index 14,241
Solubility Information Soluble in water. Insoluble in alcohol,chloroform and ether.
Physical Form Powder
Chemical Name or Material Alginic acid sodium salt, high viscosity
What is the molecular weight of Alginic Acid sodium salt, high viscosity (Cat. No. J61887.xx)?

Based on the viscosity of the chemical (1335 mPa= 1335 cps), the approximate molecular weight of Alginic Acid sodium salt, high viscosity is 120,000 Da.

Hazard Category H500
RTECSNumber AZ5820000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.