Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alginic Acid, Sodium Salt, Medium Viscosity, MP Biomedicals

Catalog No. ICN15472491 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15472491 1 kg
ICN15472480 100 g
ICN15472483 250 g
ICN15472490 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN15472491 Supplier MP Biomedicals, Inc Supplier No. 0215472491
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Source: kelp

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9005-38-3
Molecular Formula (C6H7O7)A(C6H7O7)BNa
Synonym Sodium alginate

Specifications

Color White to Cream
Physical Form Powder
Quantity 1 kg
Assay Percent Range >98%
Linear Formula (C6H7O6Na)n
Chemical Name or Material Alginic Acid, Sodium Salt, Medium Viscosity, Viscosity (1% aq. solution): 600-900 cps
Recommended Storage Room Temperature
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.