Learn More
ATHENA ES ALI (Air-Liquid Interface) Medium
Catalog No. 5000158077
Encompass_Preferred
Cell culture medium for an optimal and standardized culture of human bronchial epithelial cells at the air-liquid interface (ALI).
Catalog No. 50-001-58077 Supplier ATHENA ES Supplier No. 0458
Cell culture medium for an optimal and standardized culture of human bronchial epithelial cells at the air-liquid interface (ALI).