Learn More
Bellco Glass, Inc. ali-Q 2 VS (Variable Speed) Pipet Controller
SDP
Catalog No. 502330035
Encompass_Preferred
ali-Q 2 VS (Variable Speed) Pipet ControllerThe ali-Q 2 Series includes 3 models, varying only in the aliquoting speed/force as follows:Aliquoting speed/force:ali-Q 2 – Fixed High Speed(equal to setting 7 on VS model)ali-Q 2 LS – Fixed Low Speed(equal to setting 4 on VS model)ali-Q 2 VS – Adjustable Variable Speed(with speed setting dial)(7 speeds; 1 is slowest, 7 is fastest)Aliquoting volume range:ali-Q 2 – 0.5 – 5.0mLali-Q 2 LS – 0.3 – 3.0mLali-Q 2 VS – 0.5 – 5.0mLFeatures of the entire ali-Q 2 Series:Sensitive touch-control aspirate and dispense buttonsAliquot buttonAliquot volume dial2 multifunction LED indicatorsMultiple alert tonesRechargeable lithium ion batteryAccessories included:0.22 μm replaceable nozzle filter (1 spare)USB-C cable with chargerRemovable kickstandWall mountCalibration certificateAdditional product info:Individually serializedOne-year warrantyDimensions: 7.2″ x 7.0″ x 2.2″(18.3 cm x 17.8 cm x 5.6 cm)Weight: 350g
Catalog No. 50-233-0035 Supplier Bellco Glass, Inc. Supplier No. 451900094
ali-Q 2 VS (Variable Speed) Pipet ControllerThe ali-Q 2 Series includes 3 models, varying only in the aliquoting speed/force as follows:Aliquoting speed/force:ali-Q 2 – Fixed High Speed(equal to setting 7 on VS model)ali-Q 2 LS – Fixed Low Speed(equal to setting 4 on VS model)ali-Q 2 VS – Adjustable Variable Speed(with speed setting dial)(7 speeds; 1 is slowest, 7 is fastest)Aliquoting volume range:ali-Q 2 – 0.5 – 5.0mLali-Q 2 LS – 0.3 – 3.0mLali-Q 2 VS – 0.5 – 5.0mLFeatures of the entire ali-Q 2 Series:Sensitive touch-control aspirate and dispense buttonsAliquot buttonAliquot volume dial2 multifunction LED indicatorsMultiple alert tonesRechargeable lithium ion batteryAccessories included:0.22 μm replaceable nozzle filter (1 spare)USB-C cable with chargerRemovable kickstandWall mountCalibration certificateAdditional product info:Individually serializedOne-year warrantyDimensions: 7.2″ x 7.0″ x 2.2″(18.3 cm x 17.8 cm x 5.6 cm)Weight: 350g