Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alibendol 98.0+%, TCI America™
SDP

Catalog No. A29655G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A29655G 5 g
A29651G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A29655G Supplier TCI America Supplier No. A29655G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 26750-81-2
Molecular Formula C13H17NO4
Molecular Weight (g/mol) 251.282
MDL Number MFCD00865165
InChI Key UMJHTFHIQDEGKB-UHFFFAOYSA-N
Synonym 5-Allyl-N-(2-hydroxyethyl)-3-methoxysalicylamide, 5-Allyl-2-hydroxy-N-(2-hydroxyethyl)-3-methoxybenzamide, Cebera
PubChem CID 71916
IUPAC Name 2-hydroxy-N-(2-hydroxyethyl)-3-methoxy-5-prop-2-enylbenzamide
SMILES COC1=C(C(=CC(=C1)CC=C)C(=O)NCCO)O

Specifications

Melting Point 100°C
Color White-Yellow
Quantity 5 g
Formula Weight 251.28
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Alibendol
RTECSNumber CU8730000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.