Learn More
*LENIOBIO GMBH ALiCE for Scale-Up 10ml 1/EA
Supplier:  *LENIOBIO GMBH BLK0103011
Encompass_Preferred
Production of almost any functional protein is possible with the ALiCE cell-free protein expression system. With no optimization needed for modifications such as disulfide bonding or N-glycosylation, ALiCE brings production of membrane proteins, antibodies and protein complexes within easy reach. The platform is automation friendly and scalable – from expression tests and rapid prototyping right through to manufacture.
Catalog No. NC3404522