Learn More
LENIOBIO US CORP ALiCE for Scale-Up 25ml 1/EA
Catalog No. NC3404526
Encompass_Preferred
Production of almost any functional protein is possible with the ALiCE cell-free protein expression system. With no optimization needed for modifications such as disulfide bonding or N-glycosylation, ALiCE brings production of membrane proteins, antibodies and protein complexes within easy reach. The platform is automation friendly and scalable – from expression tests and rapid prototyping right through to manufacture.
Catalog No. NC3404526 Supplier LENIOBIO US CORP Supplier No. BLK0103026
Add to Cart
Add to Cart
Request a Quote
Production of almost any functional protein is possible with the ALiCE cell-free protein expression system. With no optimization needed for modifications such as disulfide bonding or N-glycosylation, ALiCE brings production of membrane proteins, antibodies and protein complexes within easy reach. The platform is automation friendly and scalable – from expression tests and rapid prototyping right through to manufacture.