Learn More
LENIOBIO US CORP ALiCE for Scale-Up 5ml 1/EA
Catalog No. NC3404510
Encompass_Preferred
Production of almost any functional protein is possible with the ALiCE cell-free protein expression system. With no optimization needed for modifications such as disulfide bonding or N-glycosylation, ALiCE brings production of membrane proteins, antibodies and protein complexes within easy reach. The platform is automation friendly and scalable – from expression tests and rapid prototyping right through to manufacture.
Catalog No. NC3404510 Supplier LENIOBIO US CORP Supplier No. BLK0103005
Production of almost any functional protein is possible with the ALiCE cell-free protein expression system. With no optimization needed for modifications such as disulfide bonding or N-glycosylation, ALiCE brings production of membrane proteins, antibodies and protein complexes within easy reach. The platform is automation friendly and scalable – from expression tests and rapid prototyping right through to manufacture.