Learn More
Newcomer Supply Alcian Blue/PAS Control Slides, 10/set
SDP
Catalog No. NC0779507
Encompass
Newcomer Supply Alcian Blue/PAS Control Slides are for the positive histochemical staining and differentiation of acidic epithelial mucins (sialomucin, sulfomucin), stromal (mesenchymal) mucin, neutral mucins and glycogen. Alcian Blue/PAS Control Slides contain sections of positive staining small intestine.
Catalog No. NC0779507 Supplier Newcomer Supply Supplier No. 4022A
Newcomer Supply Alcian Blue/PAS Control Slides are for the positive histochemical staining and differentiation of acidic epithelial mucins (sialomucin, sulfomucin), stromal (mesenchymal) mucin, neutral mucins and glycogen. Alcian Blue/PAS Control Slides contain sections of positive staining small intestine.