Learn More
Analytical Sales & Services Alignment Spacer for Lumidox II LED Arrays, for use with Bio-Rad HSP9601 Plates
Catalog No. 5000805821
Encompass
Plate Alignment Spacer\n\nUse to “couple” Lumidox II LED Lens Mat Array with Bio-Rad HSP9601 Plates\n\nQty/Pkg: Each
Catalog No. 50-008-05821 Supplier Analytical Sales & Services Supplier No. A132659
Plate Alignment Spacer\n\nUse to “couple” Lumidox II LED Lens Mat Array with Bio-Rad HSP9601 Plates\n\nQty/Pkg: Each