Learn More
Analytical Sales & Services Alignment Spacer for LumidoxII 24-position LED Arrays, for use with 24 Deep Well Plate #24128
Catalog No. 5000805713
Encompass
24 Deep Well Plate Alignment Spacer\n\nUse to “couple” 24 Deep Well Plate (part #24128) with Lumidox II 24-position LED Lens Mat Array\n\nQty/Pkg: Each
Catalog No. 50-008-05713 Supplier Analytical Sales & Services Supplier No. AT11028
24 Deep Well Plate Alignment Spacer\n\nUse to “couple” 24 Deep Well Plate (part #24128) with Lumidox II 24-position LED Lens Mat Array\n\nQty/Pkg: Each