Learn More
Analytical Sales & Services Alignment Spacer for LumidoxII LED Arrays, for use with 960030, 960038, 960130 and 960138 Well Plates
Catalog No. 5000805671
Encompass
Cell Culture Plate Alignment Spacer\n\nUse to “couple” Lumidox II LED Lens Mat Array with 960030, 960038, 960130 or 960138 Well Plates\n\nQty/Pkg: Each
Catalog No. 50-008-05671 Supplier Analytical Sales & Services Supplier No. AP960030
Cell Culture Plate Alignment Spacer\n\nUse to “couple” Lumidox II LED Lens Mat Array with 960030, 960038, 960130 or 960138 Well Plates\n\nQty/Pkg: Each