Learn More
Analytical Sales & Services Alignment Spacer for LumidoxII LED Arrays, for use with (but not limited to) Corning 3635 Cell Culture Plates
Catalog No. 5000805987
Encompass
Cell Culture Plate Alignment Spacer\n\n• Use to “couple” Corning Cell Culture Plates with Lumidox II LED Lens Mat Arrays\n• Compatible with (but not limited to) Corning 3635 cell culture plates
Catalog No. 50-008-05987 Supplier Analytical Sales & Services Supplier No. ACRCLP01
Cell Culture Plate Alignment Spacer\n\n• Use to “couple” Corning Cell Culture Plates with Lumidox II LED Lens Mat Arrays\n• Compatible with (but not limited to) Corning 3635 cell culture plates