Learn More
Analytical Sales & Services Alignment Spacer for LumidoxII LED Arrays, for use with Cell Culture Plate #96097 (Greiner 655097)
Catalog No. 5000805946
Encompass
Cell Culture Plate Alignment Spacer\n\n• Use to “couple” cell culture plate #96097 (Greiner 655097) with Lumidox II LED Lens Mat Array\n• Also fits Greiner 655090\n\nQty/Pkg: Each
Catalog No. 50-008-05946 Supplier Analytical Sales & Services Supplier No. A655097
Cell Culture Plate Alignment Spacer\n\n• Use to “couple” cell culture plate #96097 (Greiner 655097) with Lumidox II LED Lens Mat Array\n• Also fits Greiner 655090\n\nQty/Pkg: Each