Learn More
Analytical Sales & Services Alignment Spacer for LumidoxII LED Arrays, for use with Cell Culture Plate #961515 (Cellvis P96-15H-N)
Catalog No. 5000805915
Encompass
Cell Culture Plate Alignment Spacer\n\nUse to “couple” cell culture plate #961515 (Cellvis P96-15H-N) with Lumidox II LED Lens Mat Arrays
Catalog No. 50-008-05915 Supplier Analytical Sales & Services Supplier No. AP9615HN
Cell Culture Plate Alignment Spacer\n\nUse to “couple” cell culture plate #961515 (Cellvis P96-15H-N) with Lumidox II LED Lens Mat Arrays