Learn More
Analytical Sales & Services Alignment Spacer for LumidoxII LED Arrays, for use with Cell Culture Plate #96518 (Greiner 655180)
Catalog No. 5000805884
Encompass
Cell Culture Plate Alignment Spacer\n\nUse to "couple" 96-well, flat bottom clear microplate, part #96518 (Greiner 655180) with Lumidox II LED Lens Mat Arrays (Also compatible with Greiner 655080 and other similar plates.)
Catalog No. 50-008-05884 Supplier Analytical Sales & Services Supplier No. A655180
Cell Culture Plate Alignment Spacer\n\nUse to "couple" 96-well, flat bottom clear microplate, part #96518 (Greiner 655180) with Lumidox II LED Lens Mat Arrays (Also compatible with Greiner 655080 and other similar plates.)