Learn More
Analytical Sales & Services Alignment Spacer for LumidoxII LED Arrays, for use with Corning 2592 Plate
Catalog No. 5000805988
Encompass
Cell Culture Plate Alignment Spacer\n\nUse to “couple” Corning 2592 Plate with Lumidox II LED Lens Mat Array\n\nQty/Pkg: Each
Catalog No. 50-008-05988 Supplier Analytical Sales & Services Supplier No. ACRCLP03
Cell Culture Plate Alignment Spacer\n\nUse to “couple” Corning 2592 Plate with Lumidox II LED Lens Mat Array\n\nQty/Pkg: Each