Learn More
Analytical Sales & Services Alignment Spacer for LumidoxII LED Arrays, for use with Corning 3631 Plate
Catalog No. 5000805948
Encompass
Cell Culture Plate Alignment Spacer\n\nUse to “couple” Corning 3631 Plate with Lumidox II LED Lens Mat Array\n\nQty/Pkg: Each
Catalog No. 50-008-05948 Supplier Analytical Sales & Services Supplier No. ACRCLP02
Cell Culture Plate Alignment Spacer\n\nUse to “couple” Corning 3631 Plate with Lumidox II LED Lens Mat Array\n\nQty/Pkg: Each