Learn More
Analytical Sales & Services Alignment Spacer for LumidoxII LED Arrays, for use with Fisher plate FB012931 and ThermoFisher plate 265302
Catalog No. 5000804889
Encompass
Cell Culture Plate Alignment Spacer\n\nUse to “couple” Lumidox II LED Lens Mat Array with Fisher plate FB012931 and ThermoFisher plate 265302\n\nQty/Pkg: Each
Catalog No. 50-008-04889 Supplier Analytical Sales & Services Supplier No. AP265302
Cell Culture Plate Alignment Spacer\n\nUse to “couple” Lumidox II LED Lens Mat Array with Fisher plate FB012931 and ThermoFisher plate 265302\n\nQty/Pkg: Each