Learn More
Medchemexpress LLC Alimemazine-d6 (Trimeprazine d6) | 1346603-88-0 | 99.6% | 10 MG
SDP
Catalog No. 5000341372
Encompass_Preferred
Alimemazine-d6 is a deuterium-labeled antihistamine intended for research use. It serves as a tracer and an internal standard for quantitative analysis methods such as NMR, GC-MS, or LC-MS. This compound has a molecular weight of 304.48 and a formula of C18H16D6N2S, appearing as a white to off-white solid.
  • Deuterium-labeled antihistamine
  • Intended for research use
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis
  • Suitable for NMR, GC-MS, and LC-MS applications
  • White to off-white solid appearance
Catalog No. 50-003-41372 Supplier Medchemexpress LLC Supplier No. HY12752S10MG
Alimemazine-d6 is a deuterium-labeled antihistamine intended for research use. It serves as a tracer and an internal standard for quantitative analysis methods such as NMR, GC-MS, or LC-MS. This compound has a molecular weight of 304.48 and a formula of C18H16D6N2S, appearing as a white to off-white solid.
  • Deuterium-labeled antihistamine
  • Intended for research use
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis
  • Suitable for NMR, GC-MS, and LC-MS applications
  • White to off-white solid appearance