Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alimemazine Tartrate 98.0+%, TCI America™
SDP

Catalog No. A2813100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A2813100MG 100 mg
A28131G 1 g
A28135G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A2813100MG Supplier TCI America Supplier No. A2813100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4330-99-8
Molecular Formula C40H50N4O6S2
Molecular Weight (g/mol) 746.982
MDL Number MFCD00242594
InChI Key AJZJIYUOOJLBAU-RNKHSWPKSA-N
Synonym Trimeprazine Tartrate, 10-[3-(Dimethylamino)-2-methylpropyl]phenothiazine Tartrate
PubChem CID 45358050
IUPAC Name (2S,3S)-2,3-dihydroxybutanedioic acid;N,N,2-trimethyl-3-phenothiazin-10-ylpropan-1-amine
SMILES CC(CN1C2=CC=CC=C2SC3=CC=CC=C31)CN(C)C.CC(CN1C2=CC=CC=C2SC3=CC=CC=C31)CN(C)C.C(C(C(=O)O)O)(C(=O)O)O

Specifications

Melting Point 161°C
Color White-Yellow
Quantity 100 mg
UN Number 2811
Formula Weight 746.98
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Alimemazine Tartrate
RTECSNumber SO6475000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.