Learn More
Cayman Chemical α-Linolenic Acid ethyl ester, Purity: ≥98%, CAS: 1191-41-9, 100 mg
SDP
Catalog No. 502887597
Encompass_Preferred
An esterified form of α-linolenic acid; increases cyclin E levels and the activity of Cdk2/cyclin E, ERK, and JNK in hepatic stellate cells at 50 μM; inhibits the growth of S. mutans, C. albicans, and P. gingivalis at 25 μg/ml; has been found in biodiesel produced from castor oil using ethanol; has been used as a substrate in lipid peroxidation assays for antioxidant activity
Catalog No. 50-288-7597 Supplier Cayman Chemical Supplier No. 10008199100 MG
An esterified form of α-linolenic acid; increases cyclin E levels and the activity of Cdk2/cyclin E, ERK, and JNK in hepatic stellate cells at 50 μM; inhibits the growth of S. mutans, C. albicans, and P. gingivalis at 25 μg/ml; has been found in biodiesel produced from castor oil using ethanol; has been used as a substrate in lipid peroxidation assays for antioxidant activity