Learn More
Cayman Chemical α-Linolenic Acid (sodium salt), Purity: ≥95%, CAS: 822-18-4, 100 mg
SDP
Catalog No. 502985922
Encompass_Preferred
An essential ω-3 polyunsaturated fatty acid found in plants; converted to eicosapentaenoic, docosapentaenoic, and docosahexaenoic acids, catabolized via fatty acid β-oxidation for energy or the synthesis of saturated and monounsaturated fatty acids, or stored in adipose tissue in vivo; dietary consumption is positively associated with a moderately lower risk of cardiovascular disease
Catalog No. 50-298-5922 Supplier Cayman Chemical Supplier No. 21910100 MG
An essential ω-3 polyunsaturated fatty acid found in plants; converted to eicosapentaenoic, docosapentaenoic, and docosahexaenoic acids, catabolized via fatty acid β-oxidation for energy or the synthesis of saturated and monounsaturated fatty acids, or stored in adipose tissue in vivo; dietary consumption is positively associated with a moderately lower risk of cardiovascular disease