Learn More
Cayman Chemical aLInolenoyl EthnolamIDd4 100UG
SDP
Catalog No. 502895568
Encompass_Preferred
An internal standard for the quantification of α-linolenoyl ethanolamide by GC- or LC-MS
Catalog No. 50-289-5568 Supplier Cayman Chemical Supplier No. 9001841100 UG
An internal standard for the quantification of α-linolenoyl ethanolamide by GC- or LC-MS