Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aliquat∣r 336 TG

Catalog No. AAA172470F
Click to view available options
Quantity:
100 mL
2500 mL

CAS: 63393-96-4 | C25H54ClN | 404.16 g/mol

Aliquat 336 is widely utilized as a phase transfer catalyst for the catalytic oxidation of cyclohexene to 1,6-hexanedioic acid. It is involved in the total synthesis of Manzamine A. It serves as an anti-static agent for textile fabrics and carpeting. It finds application in waste water treatment to control foaming. It has a major application as a metal extraction reagent for cadmium, zinc, iron and rare earth metals. It is used as an effective catalyst for the preparation of methylphenyl ether from sodium phenolate with for methyl bromide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Aliquat 336 is widely utilized as a phase transfer catalyst for the catalytic oxidation of cyclohexene to 1,6-hexanedioic acid. It is involved in the total synthesis of Manzamine A. It serves as an anti-static agent for textile fabrics and carpeting. It finds application in waste water treatment to control foaming. It has a major application as a metal extraction reagent for cadmium, zinc, iron and rare earth metals. It is used as an effective catalyst for the preparation of methylphenyl ether from sodium phenolate with for methyl bromide.

Solubility
Miscible with benzene and chloroform.

Notes
Incompatible with strong oxidizing agents, acids and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63393-96-4
Molecular Formula C25H54ClN
MDL Number MFCD00011862
InChI Key XKBGEWXEAPTVCK-UHFFFAOYSA-M
Synonym methyltrioctylammonium chloride, aliquat 336, trioctylmethylammonium chloride, methyl trioctyl ammonium chloride, tomac, tricaprylmethylammonium chloride, capriquat, methyltricaprylylammonium chloride, tricaprylylmethylammonium chloride, trioctyl methyl ammonium chloride
PubChem CID 21218
ChEBI CHEBI:75286
IUPAC Name methyl(trioctyl)azanium;chloride

Specifications

Density 0.88
Boiling Point >240°C
Flash Point 132°C (269°F)
Refractive Index 1.4465
Quantity 2500 mL
UN Number UN2810
Sensitivity Hygroscopic
Solubility Information Miscible with benzene and chloroform.
SMILES [Cl-].CCCCCCCC[N+](C)(CCCCCCCC)CCCCCCCC
Molecular Weight (g/mol) 404.16
Formula Weight 404.17
Chemical Name or Material Aliquat™ 336
Show More Show Less
Hazard Category H301-H314-H335-H360FD-H373
Hazard Statement GHS H Statement
H301-H318-H315
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 264-120-7
RTECSNumber BR8575000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.