Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aliquat™ 336TG

Catalog No. AC463570025
Change view
Click to view available options
Quantity:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC463570025 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC463570025 Supplier Thermo Scientific Chemicals Supplier No. 463570025

missing translation for 'validMainframeMsg'

CAS: 63393-96-4 | C25H54ClN | 404.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63393-96-4
Molecular Formula C25H54ClN
Molecular Weight (g/mol) 404.16
InChI Key XKBGEWXEAPTVCK-UHFFFAOYSA-M
IUPAC Name methyltrioctylazanium chloride
SMILES [Cl-].CCCCCCCC[N+](C)(CCCCCCCC)CCCCCCCC

Specifications

Density 0.8900 g/mL
Flash Point 77°C
Quantity 2.5 L
Specific Gravity 0.89
Solubility Information Solubility aq. soln.: insoluble.
Viscosity 1500 mPa.s (30°C)
Chemical Name or Material Aliquat* 336 TG
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Toxic if swallowed.

May cause damage to organs through prolonged or repeated exposure.

Causes severe skin burns and eye damage.

May damage fertility.

May damage the unborn child.

Very toxic to aquatic l
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection.

IF SWALLOWED: Rinse mouth.

Do NOT induce vomiting.

IF ON SKIN (or hair): Take off immediately all contaminated clothing.

Rinse skin with water or shower IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

Immediately call a POISON CENTER or doctor/physician.

EINECSNumber 264-120-7

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.