Learn More
Medchemexpress LLC Alirocumab 10mg | 10mg
SDP
Catalog No. 5000322128
Encompass_Preferred
Alirocumab is an anti-PCSK9 human monoclonal antibody Alirocumab inhibits PCSK9 Alirocumab reduces NLRP3 inflammasome regulates Nrf2/HO-1 HMGB1/NF- B and Fractalkine/CX3CR1 Alirocumab increases the ability of the liver to bind LDL-cholesterol (LDL-C) and reduces levels of LDL-C in blood Alirocumab improves atherosclerosis and inflammation[1][2][3][4][5][6][7][8][9][10][11]
Catalog No. 50-003-22128 Supplier Medchemexpress LLC Supplier No. HYP992810MG
Alirocumab is an anti-PCSK9 human monoclonal antibody Alirocumab inhibits PCSK9 Alirocumab reduces NLRP3 inflammasome regulates Nrf2/HO-1 HMGB1/NF- B and Fractalkine/CX3CR1 Alirocumab increases the ability of the liver to bind LDL-cholesterol (LDL-C) and reduces levels of LDL-C in blood Alirocumab improves atherosclerosis and inflammation[1][2][3][4][5][6][7][8][9][10][11]