Learn More
Medchemexpress LLC HY-10971 10mg Medchemexpress, Alisertib CAS:1028486-01-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1097110MG
Encompass_Preferred
Medchemexpress, HY-10971 10mg Alisertib CAS:1028486-01-2 Alisertib (MLN 8237) is an orally active and selective Aurora A kinase inhibitor (IC50=1.2 nM), which binds to Aurora A kinase resulting in mitotic spindle abnormalities, mitotic accumulation. Alisertib (MLN 8237) induces apoptosis and autophagy through targeting the AKT/mTOR/AMPK/p38 pathway in leukemic cells. Antitumor activity[2][3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8797