Learn More
Apexbio Technology LLC Aliskiren 173334-57-1 1g
SDP
Catalog No. 5000095801
Encompass_Preferred
Aliskiren (CAS 173334-57-1) is a small-molecule inhibitor that targets human renin an aspartyl protease involved in the regulation of the renin angiotensin aldosterone system (RAAS) By directly inhibiting renin activity Aliskiren suppresses the conversion of angiotensinogen to angiotensin I thereby modulating downstream effects on blood pressure regulation It is clinically approved for the management of essential hypertension In biomedical research Aliskiren serves as a tool compound to investigate RAAS-mediated pathways and the physiological consequences of direct renin inhibition in cardiovascular renal and metabolic disease models
Catalog No. 50-000-95801 Supplier Apexbio Technology LLC Supplier No. A31531000
Aliskiren (CAS 173334-57-1) is a small-molecule inhibitor that targets human renin an aspartyl protease involved in the regulation of the renin angiotensin aldosterone system (RAAS) By directly inhibiting renin activity Aliskiren suppresses the conversion of angiotensinogen to angiotensin I thereby modulating downstream effects on blood pressure regulation It is clinically approved for the management of essential hypertension In biomedical research Aliskiren serves as a tool compound to investigate RAAS-mediated pathways and the physiological consequences of direct renin inhibition in cardiovascular renal and metabolic disease models