Learn More
Apexbio Technology LLC Aliskiren Hemifumarate 173334-58-2 100mg
SDP
Catalog No. 502240247
Encompass_Preferred
Aliskiren Hemifumarate (CAS 173334-58-2) is a small molecule classified as a direct renin inhibitor By binding to renin it blocks the initial step in the renin-angiotensin-aldosterone system (RAAS) thereby reducing the conversion of angiotensinogen to angiotensin I and lowering downstream angiotensin II levels This mechanism results in decreased vasoconstriction and aldosterone-mediated sodium retention Aliskiren Hemifumarate is primarily studied in the context of essential hypertension and serves as a tool compound for exploring renin-dependent regulatory pathways and therapeutic strategies targeting blood pressure homeostasis
This item is not returnable. View return policy
Catalog No. 50-224-0247 Supplier Apexbio Technology LLC Supplier No. B2215100
Aliskiren Hemifumarate (CAS 173334-58-2) is a small molecule classified as a direct renin inhibitor By binding to renin it blocks the initial step in the renin-angiotensin-aldosterone system (RAAS) thereby reducing the conversion of angiotensinogen to angiotensin I and lowering downstream angiotensin II levels This mechanism results in decreased vasoconstriction and aldosterone-mediated sodium retention Aliskiren Hemifumarate is primarily studied in the context of essential hypertension and serves as a tool compound for exploring renin-dependent regulatory pathways and therapeutic strategies targeting blood pressure homeostasis