Learn More
TARGETMOL CHEMICALS INC ALISKIREN HEMIFUMARATE 100MG
SDP
Catalog No. 502581245
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 50 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Aliskiren hemifumarate (CGP60536B) is an orally active nonpeptide renin inhibitor with antihypertensive activity. purity: 99%
Catalog No. 50-258-1245 Supplier TARGETMOL CHEMICALS INC Supplier No. T1520100MG
Also available in 5 mg 10 mg 25 mg 50 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Aliskiren hemifumarate (CGP60536B) is an orally active nonpeptide renin inhibitor with antihypertensive activity. purity: 99%