Learn More
Cayman Chemical AlIskIrenhemIfumarate 10mg
SDP
Catalog No. 502984224
Encompass_Preferred
A nonpeptide renin inhibitor (IC50 = 0.6 nM); reduces blood pressure and prevents albuminuria in renin-overexpressing TG(mRen-2)27 rats in a model of streptozotocin-induced diabetes at 10 and 30 mg/kg
Catalog No. 50-298-4224 Supplier Cayman Chemical Supplier No. 1964010 MG
A nonpeptide renin inhibitor (IC50 = 0.6 nM); reduces blood pressure and prevents albuminuria in renin-overexpressing TG(mRen-2)27 rats in a model of streptozotocin-induced diabetes at 10 and 30 mg/kg