Learn More
Medchemexpress LLC Alisol A 24-acetate | 18674-16-3 | 98.0% | 532.75 | C32H52O6 | 10 MG
SDP
Catalog No. 5000296195
Encompass_Preferred
Alisol A 24-acetate is a naturally occurring prototerpene-type triterpenoid supplied as a high-purity research chemical for biochemical and pharmacological studies. It is provided as a solid or as a ready-to-use DMSO solution for assay applications and is intended for research use only.
  • high purity: 98.0% by analytical specification
  • small-scale quantities suitable for screening and pilot studies
  • available as a solid and as a 10 mM solution in DMSO
  • molecular formula: C32H52O6; molecular weight: 532.75
  • provided with datasheet, certificate of analysis, and safety data sheet
Catalog No. 50-002-96195 Supplier Medchemexpress LLC Supplier No. HYN0853A10MG
Alisol A 24-acetate is a naturally occurring prototerpene-type triterpenoid supplied as a high-purity research chemical for biochemical and pharmacological studies. It is provided as a solid or as a ready-to-use DMSO solution for assay applications and is intended for research use only.
  • high purity: 98.0% by analytical specification
  • small-scale quantities suitable for screening and pilot studies
  • available as a solid and as a 10 mM solution in DMSO
  • molecular formula: C32H52O6; molecular weight: 532.75
  • provided with datasheet, certificate of analysis, and safety data sheet