Learn More
Medchemexpress LLC Alisol B | 18649-93-9 | 472.70 | 1 MG
SDP
Catalog No. 5000416278
Encompass_Preferred
Alisol B is a triterpene that exhibits diverse biological activities, making it valuable for various research applications. It is known to bind human soluble epoxide hydrolase (sEH) and reduce its activity. This compound also plays a role in inhibiting RANKL-induced JNK phosphorylation, osteoclast formation, and SERCA pump activity, while promoting calcium mobilization and autophagy. Alisol B can be used for research into conditions such as hypercalcemia, osteoporosis, rheumatoid arthritis, acute kidney injury, and breast cancer.
  • Inhibits sEH activity
  • Reduces osteoclast formation
  • Induces calcium mobilization
  • Activates CaMKK-AMPK-mTOR pathway
  • Promotes autophagic flux and autophagosome formation
  • Induces G1 phase cell cycle arrest
  • Activates endoplasmic reticulum stress and unfolded protein responses
  • Induces cancer cell apoptosis
Catalog No. 50-004-16278 Supplier Medchemexpress LLC Supplier No. HYN0805A1MG
Alisol B is a triterpene that exhibits diverse biological activities, making it valuable for various research applications. It is known to bind human soluble epoxide hydrolase (sEH) and reduce its activity. This compound also plays a role in inhibiting RANKL-induced JNK phosphorylation, osteoclast formation, and SERCA pump activity, while promoting calcium mobilization and autophagy. Alisol B can be used for research into conditions such as hypercalcemia, osteoporosis, rheumatoid arthritis, acute kidney injury, and breast cancer.
  • Inhibits sEH activity
  • Reduces osteoclast formation
  • Induces calcium mobilization
  • Activates CaMKK-AMPK-mTOR pathway
  • Promotes autophagic flux and autophagosome formation
  • Induces G1 phase cell cycle arrest
  • Activates endoplasmic reticulum stress and unfolded protein responses
  • Induces cancer cell apoptosis