Learn More
TARGETMOL CHEMICALS INC Alisol B 23-acetate 25MG
SDP
Catalog No. 502877908
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. 1. Alisol B 23-acetate (Alisol B Acetate) produces protective effect against ANIT-induced hepatotoxity and cholestasis, due to FXR-mediated regulation of transporters and enzymes. 2. Alisol B 23-acetate produces promotive effect on liver regeneration, due to FXR-mediated regulation of genes involved in hepatocyte proliferation and hepato-protection. 3. Alisol B 23-acetate produces a protective effect against CCl4-induced hepatotoxicity, due to FXR and STAT3-mediated gene regulation. Purity 99.87%
Catalog No. 50-287-7908 Supplier TARGETMOL CHEMICALS INC Supplier No. T6S224625MG
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. 1. Alisol B 23-acetate (Alisol B Acetate) produces protective effect against ANIT-induced hepatotoxity and cholestasis, due to FXR-mediated regulation of transporters and enzymes. 2. Alisol B 23-acetate produces promotive effect on liver regeneration, due to FXR-mediated regulation of genes involved in hepatocyte proliferation and hepato-protection. 3. Alisol B 23-acetate produces a protective effect against CCl4-induced hepatotoxicity, due to FXR and STAT3-mediated gene regulation. Purity 99.87%