Learn More
Medchemexpress LLC Alisol B 23-acetate | 26575-95-1 | 514.74 | 5 MG
SDP
Catalog No. 5000416756
Encompass_Preferred
Alisol B 23-acetate is an orally active prototerpane-type triterpenoid isolated from *Alisma orientalis*. It induces apoptosis, promotes reactive oxygen species (ROS) generation, and modulates key cellular pathways. This compound demonstrates anti-inflammatory and hepatoprotective activities, protects kidneys from ischemia-reperfusion injury, and shows anticancer efficacy against ovarian, colon, lung, and gastric cancers. It is also utilized in studies of atherosclerosis and allergic asthma.
  • Induces apoptosis and promotes ROS generation
  • Modulates CDK4/6, MMP-2/9, PARP, FXR, and Syk pathways
  • Exhibits anti-inflammatory and hepatoprotective activities
  • Protects kidneys from ischemia-reperfusion injury
  • Demonstrates anticancer activity against various cancers
  • Useful for studying atherosclerosis and allergic asthma
Catalog No. 50-004-16756 Supplier Medchemexpress LLC Supplier No. HYN08055MG
Alisol B 23-acetate is an orally active prototerpane-type triterpenoid isolated from *Alisma orientalis*. It induces apoptosis, promotes reactive oxygen species (ROS) generation, and modulates key cellular pathways. This compound demonstrates anti-inflammatory and hepatoprotective activities, protects kidneys from ischemia-reperfusion injury, and shows anticancer efficacy against ovarian, colon, lung, and gastric cancers. It is also utilized in studies of atherosclerosis and allergic asthma.
  • Induces apoptosis and promotes ROS generation
  • Modulates CDK4/6, MMP-2/9, PARP, FXR, and Syk pathways
  • Exhibits anti-inflammatory and hepatoprotective activities
  • Protects kidneys from ischemia-reperfusion injury
  • Demonstrates anticancer activity against various cancers
  • Useful for studying atherosclerosis and allergic asthma