Learn More
Medchemexpress LLC Alisol C 23-acetate | 26575-93-9 | 528.72 | 100 MG
SDP
Catalog No. 5000416068
Encompass_Preferred
Alisol C 23-acetate is a natural product extracted from Alisma orinentale, known for its ability to significantly inhibit delayed-type hypersensitivity (DTH) response after oral administration. It is primarily used in cardiovascular disease research.
  • Inhibits delayed-type hypersensitivity response
  • Natural product extracted from Alisma orinentale
  • Used in cardiovascular disease research
  • Classified as Triterpene and Terpenoid
Catalog No. 50-004-16068 Supplier Medchemexpress LLC Supplier No. HYN0856100MG
Alisol C 23-acetate is a natural product extracted from Alisma orinentale, known for its ability to significantly inhibit delayed-type hypersensitivity (DTH) response after oral administration. It is primarily used in cardiovascular disease research.
  • Inhibits delayed-type hypersensitivity response
  • Natural product extracted from Alisma orinentale
  • Used in cardiovascular disease research
  • Classified as Triterpene and Terpenoid