Learn More
Medchemexpress LLC Alisol C 23-acetate | 26575-93-9 | 528.72 | 25 MG
SDP
Catalog No. 5000390305
Encompass_Preferred
Alisol C 23-acetate, a natural product extracted from Alisma orientale, can significantly and strongly inhibit DTH response after oral administration.
  • High purity: 99.86%
  • White to off-white solid appearance
  • Stable under recommended storage conditions
  • Recommended storage at 4°C, protected from light
  • For research use only
Catalog No. 50-003-90305 Supplier Medchemexpress LLC Supplier No. 25MGHYN0856
Alisol C 23-acetate, a natural product extracted from Alisma orientale, can significantly and strongly inhibit DTH response after oral administration.
  • High purity: 99.86%
  • White to off-white solid appearance
  • Stable under recommended storage conditions
  • Recommended storage at 4°C, protected from light
  • For research use only