Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Alisol C 23-acetate | 26575-93-9 | 528.72 | 5 MG
SDP

Catalog No. 5000416618
Encompass_Preferred

Alisol C 23-acetate is a natural product extracted from Alisma orinentale. This compound can significantly and strongly inhibit delayed-type hypersensitivity (DTH) response after oral administration. It is intended for research use only.

  • Significantly inhibits delayed-type hypersensitivity response
  • Extracted from Alisma orinentale
  • High purity (99.86%)
  • White to off-white solid appearance
  • Molecular formula: C32H48O6
  • Molecular weight: 528.72
  • Demonstrates transactivation of FXR in HepG2 cells (EC50: 2.1 μM)
  • Soluble in DMSO (25 mg/mL)

Catalog No. 50-004-16618 Supplier Medchemexpress LLC Supplier No. HYN08565MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Alisol C 23-acetate is a natural product extracted from Alisma orinentale. This compound can significantly and strongly inhibit delayed-type hypersensitivity (DTH) response after oral administration. It is intended for research use only.

  • Significantly inhibits delayed-type hypersensitivity response
  • Extracted from Alisma orinentale
  • High purity (99.86%)
  • White to off-white solid appearance
  • Molecular formula: C32H48O6
  • Molecular weight: 528.72
  • Demonstrates transactivation of FXR in HepG2 cells (EC50: 2.1 μM)
  • Soluble in DMSO (25 mg/mL)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.