Learn More
Medchemexpress LLC Alisol C 23-acetate | 26575-93-9 | 528.72 | 50 MG
SDP
Catalog No. 5000415530
Encompass_Preferred
Alisol C 23-acetate, a natural product extracted from Alisma orinentale, can significantly and strongly inhibit DTH response after oral administration. It is for research use only and not sold to patients.
  • Cell line: HepG2
  • EC50: 2.1 μM
  • Transactivation of FXR (unknown origin) transfected in HepG2 cells co-expressing pBSEP/pGL4.74 incubated for 24 hrs by luciferase reporter gene assay.
Catalog No. 50-004-15530 Supplier Medchemexpress LLC Supplier No. HYN085650MG
Alisol C 23-acetate, a natural product extracted from Alisma orinentale, can significantly and strongly inhibit DTH response after oral administration. It is for research use only and not sold to patients.
  • Cell line: HepG2
  • EC50: 2.1 μM
  • Transactivation of FXR (unknown origin) transfected in HepG2 cells co-expressing pBSEP/pGL4.74 incubated for 24 hrs by luciferase reporter gene assay.