Learn More
Cayman Chemical Alisol C Monoacetate, Purity: ≥98%, CAS: 26575-93-9, 10 mg
SDP
Catalog No. 502911222
Encompass_Preferred
A triterpenoid with diverse biological activities; active against certain antibiotic-resistant strains of S. aureus, E. faecium, E. coli, and P. aeruginosa (MICs = 2.5-5 µg/ml) but not against other antibiotic-resistant strains of S. aureus or E. coli (MICs = >256 µg/ml); inhibits calcitriol-induced osteoclast formation in a co-culture of primary rat osteoblasts and bone marrow cells at 1-10 µM; reduces trabecular bone loss and serum levels of a variety of cytokines in an ovariectomized rat model of osteoporosis at 1 and 2 mg/kg; reduces ear edema in a mouse model of delayed-type hypersensitivity at 10 and 50 mg/kg
Catalog No. 50-291-1222 Supplier Cayman Chemical Supplier No. 3582310 MG
A triterpenoid with diverse biological activities; active against certain antibiotic-resistant strains of S. aureus, E. faecium, E. coli, and P. aeruginosa (MICs = 2.5-5 µg/ml) but not against other antibiotic-resistant strains of S. aureus or E. coli (MICs = >256 µg/ml); inhibits calcitriol-induced osteoclast formation in a co-culture of primary rat osteoblasts and bone marrow cells at 1-10 µM; reduces trabecular bone loss and serum levels of a variety of cytokines in an ovariectomized rat model of osteoporosis at 1 and 2 mg/kg; reduces ear edema in a mouse model of delayed-type hypersensitivity at 10 and 50 mg/kg