Learn More
Medchemexpress LLC Alisol F | 155521-45-2 | 488.70 | 1 ML
SDP
Catalog No. 5000415454
Encompass_Preferred
Alisol F is a triterpene isolated from *Alisma orientale* that possesses immunosuppressive and anti-virus functions. It exhibits inhibitory activity against hepatitis B virus (HBV) surface antigen (HBsAg) secretion in the HepG2.2.15 cell line with an IC50 of 0.6 μM.
  • Exhibits an EC50 of 15.1 μM in the transactivation of FXR in HepG2 cells.
  • Shows an IC50 of 0.6 μM for antiviral activity against Hepatitis B virus in human HepG2.2.15 cells.
  • Isolated from plants, Alismataceae, *Alisma orientale* (Samuel.) Juz.
  • Classified as a triterpene.
  • Appears as a white to off-white solid.
Catalog No. 50-004-15454 Supplier Medchemexpress LLC Supplier No. HYN085410MM/1ML
Alisol F is a triterpene isolated from *Alisma orientale* that possesses immunosuppressive and anti-virus functions. It exhibits inhibitory activity against hepatitis B virus (HBV) surface antigen (HBsAg) secretion in the HepG2.2.15 cell line with an IC50 of 0.6 μM.
  • Exhibits an EC50 of 15.1 μM in the transactivation of FXR in HepG2 cells.
  • Shows an IC50 of 0.6 μM for antiviral activity against Hepatitis B virus in human HepG2.2.15 cells.
  • Isolated from plants, Alismataceae, *Alisma orientale* (Samuel.) Juz.
  • Classified as a triterpene.
  • Appears as a white to off-white solid.