Learn More
Medchemexpress LLC HY-12559 1mg Medchemexpress, Alisporivir CAS:254435-95-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY125591MG
Encompass_Preferred
Medchemexpress, HY-12559 1mg Alisporivir CAS:254435-95-5 Alisporivir (Debio-025) is a cyclophilin inhibitor molecule with potent anti-hepatitis C virus (HCV) activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8892