Learn More
Medchemexpress LLC Alitretinoin | 5300-03-8 | MFCD00025732 | C20H28O2 | 25 MG
SDP
Catalog No. 5000375999
Encompass_Preferred
Alitretinoin, a vitamin A derivative, acts as a potent RAR/RXR agonist. It induces apoptosis, regulates the cell cycle, and exhibits anticancer, anti-inflammatory, and neuroprotection activities.
  • Potent RAR/RXR agonist
  • Induces apoptosis
  • Regulates cell cycle
  • Exhibits anticancer activity
  • Exhibits anti-inflammatory activity
  • Exhibits neuroprotection activities
  • Relevant for neurological disease research
  • Relevant for inflammation/immunology research
  • Relevant for cancer research
Catalog No. 50-003-75999 Supplier Medchemexpress LLC Supplier No. 25MGHY15128
Add to Cart
Add to Cart
Alitretinoin, a vitamin A derivative, acts as a potent RAR/RXR agonist. It induces apoptosis, regulates the cell cycle, and exhibits anticancer, anti-inflammatory, and neuroprotection activities.
  • Potent RAR/RXR agonist
  • Induces apoptosis
  • Regulates cell cycle
  • Exhibits anticancer activity
  • Exhibits anti-inflammatory activity
  • Exhibits neuroprotection activities
  • Relevant for neurological disease research
  • Relevant for inflammation/immunology research
  • Relevant for cancer research